ContactUsLinkComponent
Reaction Vessel Lid for CG-1968-W Style Reactors, Chemglass
Reaction Vessel Lid for CG-1968-W Style Reactors, Chemglass
# de catalogue 76333-376
Fournisseur:  Chemglass
Lid Reaction Vessel Ss 400Mm 7-Nk 400 MM
Reaction Vessel Lid for CG-1968-W Style Reactors, Chemglass
# de catalogue 76333-376
Fournisseur:  Chemglass
Spécifications
  • Description:
    Reaction vessel Lid complete, stainless steel, 400 mm, 7-neck, Large addition port
  • Cat. no.:
    76333-376
Spécifications
A propos de cet article
Reaction vessel lid is constructed of ¹/₄" thick #316 S.S. with a ¹/₈" thick PTFE gasket underneath. The only material exposed to the reaction is PTFE.
  • The flat design reduces the height over the standard glass reaction vessel lids
Lid has seven openings that accept 45/50 outer PTFE inserts. One neck is a bulk addition port is 80 mm reaction flange with o-ring groove. Inserts are secured to the lid using glass filed PTFE nuts. Lid can be used on any of our CG-1968-W style reactors.
Customers Who Bought This Also Bought