ContactUsLinkComponent
Mosquito Feeders, Large, 50 mm Ø Feeding Area, Membrane Style, GlassChemglass
Mosquito Feeders, Large, 50 mm Ø Feeding Area, Membrane Style, GlassChemglass
# de catalogue 76333-626
Fournisseur:  Chemglass
Feeder Glass Mosquito Large 50 Mm Dia
Mosquito Feeders, Large, 50 mm Ø Feeding Area, Membrane Style, GlassChemglass
# de catalogue 76333-626
Fournisseur:  Chemglass
Spécifications
  • Description:
    Glass mosquito feeder, Large, 50 mm dia. Feed area, membrane style
  • Cat. no.:
    76333-626
Spécifications
A propos de cet article
Glass mosquito feeder, Similar to CG-1835, but with a larger feeding surface.
Mosquito feeding area is 50 mm diameter. The outer chamber is for circulating 37 °C water and an inner chamber into which the blood is injected. Parafilm™ is stretched over the lower portion of the inner chamber. The assembled feeder is held on top of the screened box. Feeder has 10 mm OØ hose connections, while the blood chamber has an OØ of 56 mm where the Parafilm™ is installed.
Ordering information: PLEASE NOTE – Feeder is glass only, NOT supplied with Parafilm™ or container.
Customers Who Bought This Also Bought