Mobile Phase HPLC Cap Systems, Chemglass
Fournisseur: Chemglass
Certificats
A propos de cet article
The new and improved CG-1167 HPLC Cap system provides the safest and most economical way of storing high performance, low pressure liquid chromatography mobile phase, sparging preventing bubbles from entering the system as well as collecting sample waste.
The bottle is blue tinted plastic coated flat bottom, which provides additional safety by containing the glass fragments from accidents and allowing enough time to transfer samples or discarding safely.
Delivery: Plastic Coated Glass Reservoir (1), 3-Hole Cap Assembly (1),
Combination Inlet/Sparger Filter 2 um (1), CG-1164-N-02 Flangless Nuts for 1.8" Tubing (2), CG-1164-F-02 Flangeless Ferrule for ¹/₈" Tubing (2), CG-1166-10 Tefzel Plug (1), CG-1164-02 ¹/₈" PTFE Tubing n 10' long.
Combination Inlet/Sparger Filter 2 um (1), CG-1164-N-02 Flangless Nuts for 1.8" Tubing (2), CG-1164-F-02 Flangeless Ferrule for ¹/₈" Tubing (2), CG-1166-10 Tefzel Plug (1), CG-1164-02 ¹/₈" PTFE Tubing n 10' long.
Customers Who Bought This Also Bought