Cuvette Stirring Rods, FireflySci
Fournisseur: FIREFLYSCI, INC.
Certificats
A propos de cet article
Stirring rods for the manual stirring of samples in a 10 mm cuvette.
- Steam autoclavable at 121 °C (250 °F)
Stirring rods allow samples to be continuously spun at the bottom of a cuvette to prevent sedimentation and coagulation.
Customers Who Bought This Also Bought