Avantor® ACE® HILIC-N, Analytical, HPLC/UHPLC Columns
Fournisseur: HICHROM LIMITED
Certificats
A propos de cet article
Avantor® ACE® HILIC-N columns have been designed specifically for HILIC applications and are especially useful for the separation of polar analytes. These columns are part of a suit of 3 proprietary HILIC columns (HILIC-A, HILIC-B and HILIC-N) developed to enable chromatographers to achieve reproducible and successful HILIC separations using a systematic approach to method development.
- Proprietary polyhydroxy neutral character HILIC phase
- Compatible with all manufacturers’ HPLC/UHPLC systems
- Excellent column performance, inertness and robustness
- Simplified and structured approach to successful HILIC separations
- Available in a range of particle sizes and column dimensions including UHPLC/HPLC rated hardware formats
Column hardware: Staninless steel.
Ordering information: HILIC method development kits, HILIC method development guide and flowchart are available on request.
Spécifications
- Catalog No:
- 76395-972
- 76395-978
- 76395-890
- 76395-902
- 76395-858
- 76395-856
- 76395-854
- 76395-852
- 76395-918
- 76395-916
- 76396-086
- 76396-034
- 76395-948
- 76395-946
- 76396-098
- 76395-944
- 76396-108
- 76395-954
- 76395-952
- 76395-872
- 76395-870
- Carbon load:
- 7%
- 7%
- 7%
- 7%
- 7%
- 7%
- 7%
- 7%
- 7%
- 7%
- 7%
- 7%
- 7%
- 7%
- 7%
- 7%
- 7%
- 7%
- Endcapped:
- No
- No
- No
- No
- No
- No
- No
- No
- No
- No
- No
- No
- No
- No
- No
- No
- No
- No
- pH range:
- 2 - 7
- 2 - 7
- 2 - 7
- 2 - 7
- 2 - 7
- 2 - 7
- 2 - 7
- 2 - 7
- 2 - 7
- 2 - 7
- 2 - 7
- 2 - 7
- 2 - 7
- 2 - 7
- 2 - 7
- 2 - 7
- 2 - 7
- 2 - 7
- Surface area:
- 300 m²/g
- 300 m²/g
- 300 m²/g
- 300 m²/g
- 300 m²/g
- 300 m²/g
- 300 m²/g
- 300 m²/g
- 300 m²/g
- 300 m²/g
- 300 m²/g
- 300 m²/g
- 300 m²/g
- 300 m²/g
- 300 m²/g
- 300 m²/g
- 300 m²/g
- 300 m²/g