Avantor® ACE® Generix® Phenyl-Hexyl & Generix® SIL, Analytical HPLC Columns, 5 µm
Supplier: HICHROM LIMITED
Certificates
About this item
These high performance HPLC columns contain high purity spherical silica.
- Ultra-narrow particle size distribution for excellent column efficiency
- Sharp, highly symmetrical peaks for acidic, basic and neutral analytes
- Excellent column to column and batch to batch reproducibility
Each column is individually tested and supplied with a test chromatogram.
Specifications
- Catalog No:
- 76395-090
- 76395-088
- 76395-094
- 76395-092
- 76395-096
- 76395-100
- 76395-098
- 76395-104
- 76395-102
- 76395-108
- 76395-106
- 76395-112
- 76395-110
- 76395-116
- 76395-114
- 76395-118
- 76395-160
- 76395-158
- 76395-120
- 76395-126
- 76395-168
- 76395-124
- 76395-166
- 76395-130
- 76395-164
- 76395-128
- 76395-162
- 76395-176
- 76395-174
- 76395-172
- 76395-170
- 76395-134
- 76395-132
- 76395-136
- 76395-140
- 76395-138
- 76395-144
- 76395-178
- 76395-142
- 76395-148
- 76395-190
- 76395-146
- 76395-188
- 76395-152
- 76395-186
- 76395-150
- 76395-184
- 76395-156
- 76395-154
- 76395-076
- 76395-078
- 76395-082
- 76395-080
- 76395-086
- 76395-084
- Description:
- Phenyl-Hexyl
- Phenyl-Hexyl
- Phenyl-Hexyl
- Phenyl-Hexyl
- Phenyl-Hexyl
- Phenyl-Hexyl
- Phenyl-Hexyl
- Phenyl-Hexyl
- Phenyl-Hexyl
- Phenyl-Hexyl
- Phenyl-Hexyl
- Phenyl-Hexyl
- Phenyl-Hexyl
- Phenyl-Hexyl
- Phenyl-Hexyl
- Phenyl-Hexyl
- SIL
- SIL
- Phenyl-Hexyl
- Phenyl-Hexyl
- SIL
- Phenyl-Hexyl
- SIL
- Phenyl-Hexyl
- SIL
- Phenyl-Hexyl
- SIL
- SIL
- SIL
- SIL
- SIL
- SIL
- Phenyl-Hexyl
- SIL
- SIL
- SIL
- SIL
- SIL
- SIL
- SIL
- SIL
- SIL
- SIL
- SIL
- SIL
- SIL
- SIL
- SIL
- SIL
- Phenyl-Hexyl
- Phenyl-Hexyl
- Phenyl-Hexyl
- Phenyl-Hexyl
- Phenyl-Hexyl
- Phenyl-Hexyl
- Carbon load:
- 15.2%
- 15.2%
- 15.2%
- 15.2%
- 15.2%
- 15.2%
- 15.2%
- 15.2%
- 15.2%
- 15.2%
- 15.2%
- 15.2%
- 15.2%
- 15.2%
- 15.2%
- 15.2%
- —
- —
- 15.2%
- 15.2%
- —
- 15.2%
- —
- 15.2%
- —
- 15.2%
- —
- —
- —
- —
- —
- —
- 15.2%
- —
- —
- —
- —
- —
- —
- —
- —
- —
- —
- —
- —
- —
- —
- —
- —
- 15.2%
- 15.2%
- 15.2%
- 15.2%
- 15.2%
- 15.2%
- Endcapped:
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- No
- No
- Yes
- Yes
- No
- Yes
- No
- Yes
- No
- Yes
- No
- No
- No
- No
- No
- No
- Yes
- No
- No
- No
- No
- No
- No
- No
- No
- No
- No
- No
- No
- No
- No
- No
- No
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- pH range:
- 2 - 8
- 2 - 8
- 2 - 8
- 2 - 8
- 2 - 8
- 2 - 8
- 2 - 8
- 2 - 8
- 2 - 8
- 2 - 8
- 2 - 8
- 2 - 8
- 2 - 8
- 2 - 8
- 2 - 8
- 2 - 8
- 2 - 8
- 2 - 8
- 2 - 8
- 2 - 8
- 2 - 8
- 2 - 8
- 2 - 8
- 2 - 8
- 2 - 8
- 2 - 8
- 2 - 8
- 2 - 8
- 2 - 8
- 2 - 8
- 2 - 8
- 2 - 8
- 2 - 8
- 2 - 8
- 2 - 8
- 2 - 8
- 2 - 8
- 2 - 8
- 2 - 8
- 2 - 8
- 2 - 8
- 2 - 8
- 2 - 8
- 2 - 8
- 2 - 8
- 2 - 8
- 2 - 8
- 2 - 8
- 2 - 8
- 2 - 8
- 2 - 8
- 2 - 8
- 2 - 8
- 2 - 8
- 2 - 8
- Surface area:
- 350 m²/g
- 350 m²/g
- 350 m²/g
- 350 m²/g
- 350 m²/g
- 350 m²/g
- 350 m²/g
- 350 m²/g
- 350 m²/g
- 350 m²/g
- 350 m²/g
- 350 m²/g
- 350 m²/g
- 350 m²/g
- 350 m²/g
- 350 m²/g
- 350 m²/g
- 350 m²/g
- 350 m²/g
- 350 m²/g
- 350 m²/g
- 350 m²/g
- 350 m²/g
- 350 m²/g
- 350 m²/g
- 350 m²/g
- 350 m²/g
- 350 m²/g
- 350 m²/g
- 350 m²/g
- 350 m²/g
- 350 m²/g
- 350 m²/g
- 350 m²/g
- 350 m²/g
- 350 m²/g
- 350 m²/g
- 350 m²/g
- 350 m²/g
- 350 m²/g
- 350 m²/g
- 350 m²/g
- 350 m²/g
- 350 m²/g
- 350 m²/g
- 350 m²/g
- 350 m²/g
- 350 m²/g
- 350 m²/g
- 350 m²/g
- 350 m²/g
- 350 m²/g
- 350 m²/g
- 350 m²/g
- 350 m²/g