Avantor® ACE®, HILIC Method Development Kits, Analytical, HPLC/UHPLC Columns
Fournisseur: HICHROM LIMITED
Certificats
A propos de cet article
Avantor® ACE® HILIC method development kits offer all three Avantor® ACE® proprietary HILIC phases (HILIC-A, HILIC-B and HILIC-N) in one kit.
- Ideal for the separation and retention of polar analytes
- Compatible with all manufacturers’ HPLC/UHPLC systems
- A wide range of column lengths and internal diameters are available
- Stainlesss teel column hardware
These phases offer alternative HILIC selectivity to each other, and have been developed to enable chromatographers to achieve reproducible and successful HILIC separations using a systematic approach to method development.
Ordering information: A HILIC method development technical guide and A2 wall chart are available on request and illustrate a tried and tested approach to HILIC method development.
Spécifications
- Catalog No:
- 76397-326
- 76397-282
- 76397-324
- 76397-280
- 76397-322
- 76397-224
- 76397-180
- 76397-136
- 76397-222
- 76397-178
- 76397-134
- 76397-220
- 76397-284
- 76397-228
- 76397-140
- 76397-226
- 76397-182
- 76397-138
- 76397-296
- 76397-230
- 76397-340
- 76397-294
- 76397-338
- 76397-336
- 76397-194
- 76397-192
- 76397-298
- 76397-196
- Endcapped:
- No
- No
- No
- No
- No
- No
- No
- No
- No
- No
- No
- No
- No
- No
- No
- No
- No
- No
- No
- No
- No
- No
- No
- No
- No
- No
- No
- No
- Max. pressure:
- 1000 bar (14503.8 psi)
- 1000 bar (14503.8 psi)
- 1000 bar (14503.8 psi)
- 1000 bar (14503.8 psi)
- 1000 bar (14503.8 psi)
- 1000 bar (14503.8 psi)
- 1000 bar (14503.8 psi)
- 1000 bar (14503.8 psi)
- 1000 bar (14503.8 psi)
- 1000 bar (14503.8 psi)
- 1000 bar (14503.8 psi)
- 1000 bar (14503.8 psi)
- 1000 bar (14503.8 psi)
- 1000 bar (14503.8 psi)
- 1000 bar (14503.8 psi)
- 1000 bar (14503.8 psi)
- 1000 bar (14503.8 psi)
- 1000 bar (14503.8 psi)
- 1000 bar (14503.8 psi)
- 1000 bar (14503.8 psi)
- 1000 bar (14503.8 psi)
- 1000 bar (14503.8 psi)
- 1000 bar (14503.8 psi)
- 1000 bar (14503.8 psi)
- 1000 bar (14503.8 psi)
- 1000 bar (14503.8 psi)
- 1000 bar (14503.8 psi)
- 1000 bar (14503.8 psi)
- pH range:
- 2 - 7
- 2 - 7
- 2 - 7
- 2 - 7
- 2 - 7
- 2 - 7
- 2 - 7
- 2 - 7
- 2 - 7
- 2 - 7
- 2 - 7
- 2 - 7
- 2 - 7
- 2 - 7
- 2 - 7
- 2 - 7
- 2 - 7
- 2 - 7
- 2 - 7
- 2 - 7
- 2 - 7
- 2 - 7
- 2 - 7
- 2 - 7
- 2 - 7
- 2 - 7
- 2 - 7
- 2 - 7
- Surface area:
- 300 m²/g
- 300 m²/g
- 300 m²/g
- 300 m²/g
- 300 m²/g
- 300 m²/g
- 300 m²/g
- 300 m²/g
- 300 m²/g
- 300 m²/g
- 300 m²/g
- 300 m²/g
- 300 m²/g
- 300 m²/g
- 300 m²/g
- 300 m²/g
- 300 m²/g
- 300 m²/g
- 300 m²/g
- 300 m²/g
- 300 m²/g
- 300 m²/g
- 300 m²/g
- 300 m²/g
- 300 m²/g
- 300 m²/g
- 300 m²/g
- 300 m²/g