Avantor® ACE® CN-300, HPLC Columns, 3 µm
Fournisseur: HICHROM LIMITED
Certificats
A propos de cet article
These Avantor® ACE® CN-300 columns provide great reproducibility and column lifetime. These stainless steel columns are available in a wide range of particle sizes and dimensions from capillary to preparative.
- Ultra high purity, base deactivated silica with exceptional reproducibility
- Capillary to preparative dimensions
- Wide range of particle sizes (3, 5 and 10 µm)
- Designed for peptide and protein separations
- Method validation kits contain three different batches of the same phase - available on request
These columns are designed for a wide range of chromatographic applications, to provide great performance with acidic, basic and neutral molecules.
Spécifications
- Catalog No:
- 76389-922
- Carbon load:
- 2.6%
- Endcapped:
- Yes
- Max. pressure:
- 276 bar
- pH range:
- 2 - 7
- Surface area:
- 100 m²/g
- USP code:
- L10