Rebel Hybrid Microscope, Upright and Inverted, Discover Echo
Fournisseur: DISCOVER ECHO INC.
Certificats
A propos de cet article
Rebel is designed around the user to challenge the status quo. Designed from the ground up to improve microscopy.
- ECHO Pro: powerful yet simple software reimagined to collaboratively view, capture, and share images
- Ultra high resolution 13" touch display
- CMOS color camera (8 MP)
- Plan Achromat and Apochromat options available
- XY mechanical stage option available
- Ergonomic biosafety cabinet and adjustable height table options available
Versatility: Hybrid microscope easily converts between upright and inverted configurations. Brightfield and phase contrast modes available. Eyepiece option available as well.
Productivity: Our award-winning touch interface is incredibly easy to use. Eyepieces have been replaced with a 13" iPad with Retina Display to view and capture stunning crystal-clear images.
Rebel True Ergo Table and Rebel Logic + Biosafety cabinet by Labconco bring the microscope stage down flush to the tabletop, setting a new precendent in microscope ergonomics.
Ordering information: Microscope includes a one-year warranty on electrical, and five-year warranty on mechanical parts and optics. Contact your Avantor representative to demo the the newest innovations in microscopy. A Rebel Hybrid Microscope Bundle option is also available (see 76359-898).
Delivery: Includes microscope body, iPad with locking stand, 8mp CMOS color camera, power cord, microscope cover, and lens cloth.
Customers Who Bought This Also Bought