Avantor® ACE® Generix®, Analytical UHPLC Columns, 1.8 µm
Supplier: HICHROM LIMITED
Certificates
About this item
These high performance UHPLC columns contain high purity spherical silica.
- Ultra-narrow particle size distribution for excellent column efficiency
- Sharp, highly symmetrical peaks for acidic, basic and neutral analytes
- Excellent column to column and batch to batch reproducibility
Each column is individually tested and supplied with a test chromatogram.
Specifications
- Catalog No:
- 76394-694
- 76394-692
- 76394-696
- 76394-700
- 76394-698
- 76394-704
- 76394-702
- 76394-708
- 76395-060
- 76394-706
- 76394-712
- 76394-710
- 76395-062
- 76395-196
- 76394-716
- 76394-714
- 76394-722
- 76394-720
- 76395-202
- 76394-724
- 76394-690
- 76395-208
- 76394-718
- Carbon load:
- 20.3%
- 20.3%
- 20.3%
- 20.3%
- 20.3%
- 11.9%
- 11.9%
- 11.9%
- 11.9%
- 11.9%
- 11.9%
- 11.9%
- 11.9%
- 15.2%
- 15.2%
- 15.2%
- 15.2%
- 15.2%
- 20.3%
- 15.2%
- Endcapped:
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- pH range:
- 2 - 8
- 2 - 8
- 2 - 8
- 2 - 8
- 2 - 8
- 2 - 8
- 2 - 8
- 2 - 8
- 2 - 8
- 2 - 8
- 2 - 8
- 2 - 8
- 2 - 8
- 2 - 8
- 2 - 8
- 2 - 8
- 2 - 8
- 2 - 8
- 2 - 8
- 2 - 8
- Surface area:
- 350 m²/g
- 350 m²/g
- 350 m²/g
- 350 m²/g
- 350 m²/g
- 350 m²/g
- 350 m²/g
- 350 m²/g
- 350 m²/g
- 350 m²/g
- 350 m²/g
- 350 m²/g
- 350 m²/g
- 350 m²/g
- 350 m²/g
- 350 m²/g
- 350 m²/g
- 350 m²/g
- 350 m²/g
- 350 m²/g
- USP code:
- L1
- L1
- L1
- L1
- L1
- L7
- L7
- L7
- L7
- L7
- L7
- L7
- L7
- L3
- L3
- L3
- L3
- L3
- L1
- L3