Laser and Inkjet Glossy Paper Labels, GA International
Fournisseur: GA INTERNATIONAL INC
Certificats
A propos de cet article
General use permanent paper labels, for laser and inkjet desktop printers.
- Easy to customize, with free word templates available
- Permanent adhesive ensures labels won’t wrinkle or peel off
- For use with all common brands of laser and inkjet desktop printers
- Labels won’t jam printers
- Resistant to gamma irradiation (tested to up to 50 kGy)
Glossy finish labels will adhere to a variety of surfaces, including paper, glass, cardboard, and plastic. Ideal for office use on files, folders, shelves, drawers, boxes, and other containers. Provided in sheet format, the laser and inkjet printable labels will not jam printers, and can be easily customized using free online templates.
Customers Who Bought This Also Bought