Biohazard Warning Labels, GA International
Fournisseur: Avantor
Certificats
A propos de cet article
Preprinted biohazard warning labels, available on a variety of brightly colored backgrounds.
- Special construct protects against abrasion and chemical exposure
- Labels withstand a wide temperature range (–196 °C/–320 °F to 110 °C/230 °F)
- Resistant to gamma irradiation (tested to up to 50 kGy)
Specially coated, these durable labels can resist abrasion, and exposure to harsh chemicals and solvents. These warning labels are waterproof and tearproof, and able to withstand a wide range of temperatures. They are perfect for use on plastic and cardboard boxes, as well as plastic bags and tubes, in extreme environments, from long-term storage in liquid nitrogen, and multiple freeze-thaw cycles, to boiling water. Resistant to UV rays and gamma irradiation.
Customers Who Bought This Also Bought