Standard Use Piggyback Labels, GA International
Fournisseur: GA INTERNATIONAL INC
Certificats
A propos de cet article
Permanent standard use, thermal-transfer, piggyback paper labels.
- Perfect for organizing and managing records
- Multi-layer piggyback design facilitates product tracking and streamlining
- Printable with all major thermal-transfer printer brands
The labels have a label-on-label, multi-layer design that allows multiple related labels to be printed at once and placed in one location until the smaller labels within are needed to identify subsequent items. Thermal transfer printable, they are ideal for general use in research laboratories, hospitals, and clinical trial facilities, as well as warehouses and shipping/sorting facilities.
Customers Who Bought This Also Bought