Avantor® ACE® Excel® C8, HPLC/UHPLC Columns, Analytical, 3 µm
Fournisseur: HICHROM LIMITED
Certificats
A propos de cet article
These ultra-inert Avantor® ACE® Excel® C8 columns provide great reproducibility and column lifetime. These stainless steel columns are available in a wide range of particle sizes and dimensions from capillary to preparative.
- Ultra high purity, base deactivated silica with exceptional reproducibility
- Ideal for a wide range of analytes differing in hydrophobicity
- Can be used as general purpose starting point column
- Wide range of particle sizes (2, 3 and 5 µm) and complementary phases
These columns are designed for a wide range of chromatographic applications, to provide great performance with acidic, basic and neutral molecules.
Ordering information: Method validation kits contain three different batches of the same phase - available on request.
Spécifications
- Catalog No:
- 76393-380
- 76393-384
- 76393-382
- 76393-388
- 76393-386
- 76393-390
- 76393-420
- 76393-418
- 76393-396
- 76384-572
- 76393-394
- 76384-574
- 76393-402
- 76393-400
- 76393-408
- 76393-398
- Carbon load:
- 9%
- 9%
- 9%
- 9%
- 9%
- 9%
- 9%
- 9%
- 9%
- 9%
- 9%
- 9%
- 9%
- 9%
- Endcapped:
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Max. pressure:
- 1000 bar
- 1000 bar
- 1000 bar
- 1000 bar
- 1000 bar
- 1000 bar
- 1000 bar
- 1000 bar
- 1000 bar
- 1000 bar
- 1000 bar
- 1000 bar
- 1000 bar
- 1000 bar
- pH range:
- 2 - 8
- 2 - 8
- 2 - 8
- 2 - 8
- 2 - 8
- 2 - 8
- 2 - 8
- 2 - 8
- 2 - 8
- 2 - 8
- 2 - 8
- 2 - 8
- 2 - 8
- 2 - 8
- Surface area:
- 300 m²/g
- 300 m²/g
- 300 m²/g
- 300 m²/g
- 300 m²/g
- 300 m²/g
- 300 m²/g
- 300 m²/g
- 300 m²/g
- 300 m²/g
- 300 m²/g
- 300 m²/g
- 300 m²/g
- 300 m²/g
- USP code:
- L7
- L7
- L7
- L7
- L7
- L7
- L7
- L7
- L7
- L7
- L7
- L7
- L7
- L7