Thermal Transfer Repositionable Paper Labels, GA International
Fournisseur: GA INTERNATIONAL INC
Certificats
A propos de cet article
Repositionable Paper Labels are available in a wide assortment of colors to facilitate color-coding and easy identification.
- Repositionable adhesive
- Compatible with desktop thermal printers, or industrial thermal printers
- Available in 1 or 3" core sizes
General-purpose repositionable labels are supplied perforated, and available in white, yellow, orange, red, blue, green apple, pink, lavender, or silver.
Customers Who Bought This Also Bought