Cryo-OmniTAG™, Transparent Cryogenic Labels for Thermal-Transfer Printers, GA International
Fournisseur: GA INTERNATIONAL INC
Certificats
A propos de cet article
Transparent thermoplastic cryogenic labels for bio-medical research and clinical laboratories.
- Labels can withstand a wide temperature range: extreme low (–196 °C) to elevated (100 °C)
- Transparent design ensures contents of labeled containers remain visible
- Printable with all major thermal-transfer printer brands
Labels will withstand long-term storage in deep-freeze and harsh laboratory environments, including liquid and vapor phase liquid nitrogen at –196 °C (–320.8 °F), –80 °C (–112 °F) freezers, and dry ice transportation.
Customers Who Bought This Also Bought