Paper Labels for Thermal-Transfer Printers, GA International
Fournisseur: GA INTERNATIONAL INC
Certificats
A propos de cet article
For general office and laboratory organization, warehouses, and shipping facilities.
- Strong adhesive ensures labels won’t wrinkle or flag
- Available in a wide range of colors, for color-coding and easy identification
- Printable with all major thermal transfer barcode printers
- Resistant to gamma irradiation (tested to up to 50 kGy)
General use paper labels for thermal-transfer printers are coated with a permanent adhesive and are available in a range of colors. They are ideal for use as shipping labels, color-coding, price tagging, warehouses, logistics, and sorting facilities, as well as for general office and laboratory organization.
Customers Who Bought This Also Bought