Avantor® ACE®, UHPLC Column Connectors
Fournisseur: HICHROM LIMITED
Certificats
A propos de cet article
Stainless steel/ PEEK UHPLC column inlet/ outlet connectors. These column connectors are designed to connect UHPLC columns to UHPLC systems and make other similar connections. They are suitable for use with ¹/₁₆" OD tubing and virtually all manufacturers' UHPLC systems and all brands of UHPLC columns.
- Pressure rated to 1700 bar (>25000 psi)
- Usable at elevated temperatures (up to 100 °C)
- Reusable, non-permanent swaging design
- Ten make and break cycles
For optimum connector lifetime and to avoid accidental permanent swaging of the connector to the tubing by over tightening, the use of an Avantor® ACE® torque wrench is recommended.
Customers Who Bought This Also Bought