Avantor® ACE® Excel® C18-Amide, HPLC/UHPLC Columns, 2 µm
Fournisseur: HICHROM LIMITED
Certificats
A propos de cet article
Avantor® ACE® Excel® C18-Amide is a uniquely designed stationary phase offering improved retention of polar compounds, enhanced phenolic resolution and alternative selectivity to C18 bonded columns. These stainless steel columns are available in a wide range of particle sizes and dimensions from capillary to preparative.
- Ultra-high purity, base deactivated silica with exceptional reproducibility
- Ideal for a wide range of analytes differing in hydrophobicity
- Wide range of particle sizes (1.7 to 5 µm) and complementary phases
- Low bleed for UV and LC/MS compatibility
- C18 hydrophobic plus Amide interactions, compatible with 100% aqueous condition
The special design of the Avantor® ACE® C18-Amide ligand incorporates an extended alkyl chain spacer between the silica surface and amide polar group, which gives extra phase stability.
Ordering information: Method validation kits contain three different batches of the same phase - available on request.
Spécifications
- Catalog No:
- 76392-644
- 76408-408
- 76392-612
- 76392-616
- 76392-614
- 76392-620
- 76392-618
- 76408-406
- 76392-646
- 76392-624
- 76392-622
- 76392-630
- 76392-634
- 76392-632
- 76392-642
- 76392-626
- Carbon load:
- 16.4%
- 16.4%
- 16.4%
- 16.4%
- 16.4%
- 16.4%
- 16.4%
- 16.4%
- 16.4%
- 16.4%
- 16.4%
- 16.4%
- 16.4%
- 16.4%
- Endcapped:
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Max. pressure:
- 1000 bar (14504 psi)
- 1000 bar (14504 psi)
- 1000 bar (14504 psi)
- 1000 bar (14504 psi)
- 1000 bar (14504 psi)
- 1000 bar (14504 psi)
- 1000 bar (14504 psi)
- 1000 bar (14504 psi)
- 1000 bar (14504 psi)
- 1000 bar (14504 psi)
- 1000 bar (14504 psi)
- 1000 bar (14504 psi)
- 1000 bar (14504 psi)
- 1000 bar (14504 psi)
- pH range:
- 2 - 8
- 2 - 8
- 2 - 8
- 2 - 8
- 2 - 8
- 2 - 8
- 2 - 8
- 2 - 8
- 2 - 8
- 2 - 8
- 2 - 8
- 2 - 8
- 2 - 8
- 2 - 8
- Surface area:
- 300 m²/g
- 300 m²/g
- 300 m²/g
- 300 m²/g
- 300 m²/g
- 300 m²/g
- 300 m²/g
- 300 m²/g
- 300 m²/g
- 300 m²/g
- 300 m²/g
- 300 m²/g
- 300 m²/g
- 300 m²/g
- USP code:
- L1
- L1
- L1
- L1
- L1
- L1
- L1
- L1
- L1
- L1
- L1
- L1
- L1
- L1