Avantor® ACE®, HPLC Column Connectors, Finger-Tight
Fournisseur: HICHROM LIMITED
Certificats
A propos de cet article
PEEK HPLC column inlet / outlet connectors. These column connectors are designed for use at both the column inlet and outlet ends. They are suitable for use with ¹/₁₆" OD tubing and all manufacturers' HPLC systems and all brands of HPLC columns.
- Universal fitting for HPLC column connections
- Pressure rated to 350 bar (5000 psi)
- Reusable
- Inert and biocompatible, excellent chemical resistance
Customers Who Bought This Also Bought