Avantor® ACE® Excel® AQ, UHPLC Columns, 5 µm
Supplier: Avantor
Certificates
About this item
Avantor® ACE® AQ is an ultra-inert phase designed for the reversed-phase separation of very polar compounds with up to 100% aqueous eluent. ACE AQ is ideal for use with fast gradients due to its rapid re-equilibration properties and resistance to retention loss.
- Ultra high purity, base deactivated silica with exceptional reproducibility
- Retains polar compounds in 100% aqueous mobile phase
- Unique C18 bonded phase with integral polar functionality
- Ideal for use with fast gradients due to its rapid re-equilibration properties and resistance to retention loss
- Wide range of particle sizes (2 to 5 µm) and complementary phase
These columns are designed for a wide range of chromatographic applications, to provide great performance with acidic, basic and neutral molecules.
Ordering information: Method validation kits contain three different batches of the same phase - available on request.
Specifications
- Catalog No:
- 76394-268
- 76394-270
- 76394-298
- 76394-276
- 76394-280
- 76394-288
- 76394-294
- 76394-292
- Carbon load:
- 14.0%
- 14.0%
- 14.0%
- 14.0%
- 14.0%
- 14.0%
- 14.0%
- 14.0%
- Endcapped:
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Max. pressure:
- 1000 bar (14503 psi)
- 1000 bar (14503 psi)
- 1000 bar (14503 psi)
- 1000 bar (14503 psi)
- 1000 bar (14503 psi)
- 1000 bar (14503 psi)
- 1000 bar (14503 psi)
- 1000 bar (14503 psi)
- pH range:
- 2 - 8
- 2 - 8
- 2 - 8
- 2 - 8
- 2 - 8
- 2 - 8
- 2 - 8
- 2 - 8
- Surface area:
- 300 m²/g
- 300 m²/g
- 300 m²/g
- 300 m²/g
- 300 m²/g
- 300 m²/g
- 300 m²/g
- 300 m²/g
- USP code:
- L1
- L1
- L1
- L1
- L1
- L1
- L1
- L1