Avantor® ACE®, HPLC Precolumn Filters
Fournisseur: HICHROM LIMITED
Certificats
A propos de cet article
These precolumn filters with PEEK body and stainless steel frit provide protection for analytical and microbore columns. They connect directly to the column without the need for an additional holder.
- Universal filters, suitable for all HPLC columns
- Pressure rated to 350 bar (5000 psi)
- Reusable
Analytical filters are available with 0.5 and 2 µm stainless steel frit options (recommended for 3 and 5 µm columns respectively). Microbore filters have a 0.5 µm stainless steel frit and offer lower dispersion than analytical filters. They are recommended for smaller volume (shorter, narrower ID) columns.
Customers Who Bought This Also Bought