Avantor® ACE®, Preparative Column Coupler
# de catalogue 76392-066
Fournisseur: HICHROM LIMITED
Spécifications
- Description:Column coupler (for use with H0009)
- Cat. no.:76392-066
Spécifications
A propos de cet article
Column coupler made from stainless steel/PEEK suitable for connecting preparative columns to stand-alone guard cartridge holder, or for making similar HPLC connections.
Ideally suited to connect 30 mm id preparative columns to H0009 cartridge holder.
Customers Who Bought This Also Bought