Avantor® ACE®, Capillary and Nano Column Coupler
# de catalogue 76392-068
Fournisseur: HICHROM LIMITED
Spécifications
- Description:Capillary column coupler
- Cat. no.:76392-068
Spécifications
A propos de cet article
Avantor® ACE® capillary/nano column couplers are suitable for connecting capillary and nano LC columns to stand-alone capillary/nano guard cartridges or for making similar LC connections.
Customers Who Bought This Also Bought