Avantor® ACE®, UHPLC Pre-Column Filters
Fournisseur: HICHROM LIMITED
Certificats
A propos de cet article
Avantor® ACE® UHPLC pre-column filters made from stainless steel/PEEK provide protection for all UHPLC columns. They offer ultra-low dispersion for maximum compatibility with UHPLC instruments and columns. For use with Waters 'Classic' Acquity® systems, an alternative Waters Acquity® compatible version provides improved performance.
- Universal filter, suitable for all UHPLC columns
- Compatible with all UHPLC systems
- Pressure rated to 1000 bar (15000 psi)
- Reusable
Customers Who Bought This Also Bought