Avantor® ACE® C18, HPLC Columns, 2 µm, Microbore
Fournisseur: HICHROM LIMITED
Certificats
A propos de cet article
Avantor® ACE® C18 columns provide great reproducibility and column lifetime. These stainless steel columns are available in a wide range of particle sizes and dimensions from capillary to preparative. These columns are designed for a wide range of chromatographic applications, to provide great performance with acidic, basic and neutral molecules.
- Ultra high purity, base deactivated silica with exceptional reproducibility
- Ideal for a wide range of analytes differing in hydrophobicity
- Wide range of particle sizes (2 to 10 µm) and complementary phases
Maximum recommended pH: 2.0 to 8.0
Max. pressure: 400 bar (5801.5 psi).
Spécifications
- Catalog No:
- 76383-578
- 76383-508
- Carbon load:
- 15.5%
- 15.5%
- Endcapped:
- Yes
- Yes
- Max. pressure:
- 400 bar (5801.5 psi)
- 400 bar (5801.5 psi)
- pH range:
- 2 - 8
- 2 - 8
- Surface area:
- 300 m²/g
- 300 m²/g
- USP code:
- L1
- L1