Avantor® Partisphere C18, HPLC Columns, 5 µm
Fournisseur: Avantor
Certificats
A propos de cet article
Avantor® Partisphere® was one of the first commercially available spherical silicas and continues to provide reproducible, high efficiency separations.
- Spherical 5 µm porous silica
- Unique void sealing cartridge hardware
- Convenient and easy to use - hand tightened fittings
- Increased column lifetime
Stainless steel column hardware.
Customers Who Bought This Also Bought