Avantor® Ultrasphere® Silica (Si), HPLC Columns
Fournisseur: HICHROM LIMITED
Certificats
A propos de cet article
Avantor® Ultrasphere® columns remain widely referenced within both industry and academia and are recognized to provide excellent chromatographic performance.
- 3 and 5 μm spherical porous silica
- High reproducibility, narrow particle size distribution
- Optimal surface coverage for long column lifetime
Spécifications
- Catalog No:
- 76398-128
- 76398-096
- 76398-098
- 76398-100
- Endcapped:
- N/A
- N/A
- N/A
- N/A