Avantor® Ultrasphere® Cyano (CN), HPLC Columns
Fournisseur: HICHROM LIMITED
Certificats
A propos de cet article
Avantor® Ultrasphere® columns remain widely referenced within both industry and academia and are recognized to provide excellent chromatographic performance.
- 3 and 5 μm spherical porous silica
- High reproducibility, narrow particle size distribution
- Optimal surface coverage for long column lifetime
Spécifications
- Catalog No:
- 76398-126
- 76398-170
- 76398-168
- Endcapped:
- No
- No
- No