LiChrosorb® RP-8, HPLC Columns
Fournisseur: HICHROM LIMITED
Certificats
A propos de cet article
These columns with high purity base deactivated porous silica are ideal for the separation of basic molecules.
- Irregular porous silica
- High efficiency and complimentary phase selectivity
Spécifications
- Catalog No:
- 76400-232
- 76400-234
- 76400-236
- 76400-270
- 76400-272
- 76400-228
- 76400-216
- 76400-264
- 76400-220
- 76400-252
- 76400-258
- 76400-250
- 76400-238
- Carbon load:
- 9.5%
- 9.5%
- 9.5%
- 9.5%
- 9.5%
- 9.5%
- 9.5%
- 9.5%
- 9.5%
- 9.5%
- 9.5%
- 9.5%
- 9.5%
- Endcapped:
- No
- No
- No
- No
- No
- No
- No
- No
- No
- No
- No
- No
- No
- Surface area:
- 300 m²/g
- 300 m²/g
- 300 m²/g
- 300 m²/g
- 300 m²/g
- 300 m²/g
- 300 m²/g
- 300 m²/g
- 300 m²/g
- 300 m²/g
- 300 m²/g
- 300 m²/g
- 300 m²/g