Exsil ODS1, HPLC Columns
Fournisseur: HICHROM LIMITED
Certificats
A propos de cet article
Exsil HPLC columns are a useful alternative to the classical spherical HPLC silicas.
- Spherical porous silica
- Competitively priced
- Hichrom high efficiency
Exsil 100, the standard material, is supplied in a range of particle sizes and bonded phases to cover a wide range of applications.
Spécifications
- Catalog No:
- 76398-800
- 76398-802
- Carbon load:
- 7%
- 7%
- Endcapped:
- No
- No
- Surface area:
- 200 m²/g
- 200 m²/g