LiChrosorb® RP-18, HPLC Columns
Fournisseur: HICHROM LIMITED
Certificats
A propos de cet article
These columns with high purity base deactivated porous silica are ideal for the separation of basic molecules.
- Irregular porous silica
- High efficiency and complimentary phase selectivity
Spécifications
- Catalog No:
- 76400-190
- 76400-182
- 76400-184
- 76400-186
- 76400-172
- 76400-176
- 76400-178
- 76400-144
- 76400-146
- 76400-170
- 76400-202
- 76400-158
- 76400-160
- 76400-208
- 76400-152
- 76400-154
- 76400-200
- 76400-168
- Carbon load:
- 16.2%
- 16.2%
- 16.2%
- 16.2%
- 16.2%
- 16.2%
- 16.2%
- 16.2%
- 16.2%
- 16.2%
- 16.2%
- 16.2%
- 16.2%
- 16.2%
- 16.2%
- 16.2%
- 16.2%
- 16.2%
- Endcapped:
- No
- No
- No
- No
- No
- No
- No
- No
- No
- No
- No
- No
- No
- No
- No
- No
- No
- No
- Surface area:
- 300 m²/g
- 300 m²/g
- 300 m²/g
- 300 m²/g
- 300 m²/g
- 300 m²/g
- 300 m²/g
- 300 m²/g
- 300 m²/g
- 300 m²/g
- 300 m²/g
- 300 m²/g
- 300 m²/g
- 300 m²/g
- 300 m²/g
- 300 m²/g
- 300 m²/g
- 300 m²/g