LiChrospher® RP-18, HPLC Columns
Fournisseur: HICHROM LIMITED
Certificats
A propos de cet article
These LiChrospher® columns are silica available in non-endcapped C18 phases. This feature enhances the chromatography selectivity options.
- Spherical porous silica
- High efficiency packed columns
Stainless steel column hardware.
Spécifications
- Catalog No:
- 76400-578
- 76400-582
- 76400-572
- 76400-574
- 76400-548
- 76400-584
- 76400-588
- 76400-546
- 76400-560
- 76400-590
- 76400-592
- 76400-550
- 76400-552
- 76400-596
- 76400-566
- 76400-568
- Carbon load:
- 21.0%
- 21.0%
- 21.0%
- 21.0%
- 21.0%
- 21.0%
- 21.0%
- 21.0%
- 21.0%
- 21.0%
- 21.0%
- 21.0%
- 21.0%
- 21.0%
- 21.0%
- 21.0%
- Endcapped:
- No
- No
- No
- No
- No
- No
- No
- No
- No
- No
- No
- No
- No
- No
- No
- No
- Surface area:
- 350 m²/g
- 350 m²/g
- 350 m²/g
- 350 m²/g
- 350 m²/g
- 350 m²/g
- 350 m²/g
- 350 m²/g
- 350 m²/g
- 350 m²/g
- 350 m²/g
- 350 m²/g
- 350 m²/g
- 350 m²/g
- 350 m²/g
- 350 m²/g