LiChrospher® Diol, HPLC Columns
Fournisseur: HICHROM LIMITED
Certificats
A propos de cet article
LiChrospher® is a silica available in both endcapped and non-endcapped C8 and C18 phases.
- Spherical porous silica
- Base deactivated LiChrospher® RP-select B
- High efficiency packed columns
Column hardware: Stainless steel.
Spécifications
- Catalog No:
- 76400-520
- 76400-522
- 76400-514
- 76400-504
- 76400-540
- 76400-496
- 76400-498
- 76400-500
- 76400-490
- 76400-524
- 76400-526
- 76400-528
- Carbon load:
- 8.0%
- 8.0%
- 8.0%
- 8.0%
- 8.0%
- 8.0%
- 8.0%
- 8.0%
- 8.0%
- 8.0%
- 8.0%
- 8.0%
- Endcapped:
- No
- No
- No
- No
- No
- No
- No
- No
- No
- No
- No
- No
- Surface area:
- 350 m²/g
- 350 m²/g
- 350 m²/g
- 350 m²/g
- 350 m²/g
- 350 m²/g
- 350 m²/g
- 350 m²/g
- 350 m²/g
- 350 m²/g
- 350 m²/g
- 350 m²/g