LiChrospher® CN, HPLC Columns
Fournisseur: Avantor
Certificats
A propos de cet article
LiChrospher® is a silica available in both endcapped and non-endcapped C8 and C18 phases.
- Spherical porous silica
- Base deactivated LiChrospher® RP-select B
- High efficiency packed columns
Column hardware: Stainless steel.
Spécifications
- Catalog No:
- 76400-400
- 76400-416
- 76400-418
- Carbon load:
- 6.6%
- 6.6%
- 6.6%
- Endcapped:
- No
- No
- No
- Surface area:
- 350 m²/g
- 350 m²/g
- 350 m²/g