Collapsible Ice Buckets, 5 Liters
# de catalogue 470324-996
Fournisseur: MTC BIO, INC.
Spécifications
- Description:Collapsible ice bucket
- Cat. no.:470324-996
Spécifications
A propos de cet article
This collapsible ice bucket is ideal for any lab looking to conserve storage space.
- Inner-lid access port for better temperature retention
- Attached handle for easy transport
- Molded grip on base for pouring
- Collapsible body saves storage space
BioBucket can be used as an ice bucket to keep samples cool over long periods of time, or as portable liquid container. The attached handle makes transportation a breeze while the molded base grip allows for easy pouring and cleaning. It is perfect for field use as an environmental sample collection and transport receptacle.
Customers Who Bought This Also Bought