LiChrospher® Si 60, HPLC Columns
Fournisseur: Avantor
Certificats
A propos de cet article
LiChrospher® is a silica available in both endcapped and non-endcapped C8 and C18 phases.
- Spherical porous silica
- Base deactivated LiChrospher® RP-select B
- High efficiency packed columns
Column hardware: Stainless steel.
Spécifications
- Catalog No:
- 76400-370
- 76400-346
- 76400-358
- 76400-352
- 76400-366
- Surface area:
- 700 m²/g
- 700 m²/g
- 700 m²/g
- 700 m²/g
- 700 m²/g