SW® TrueForm® TF-95RB Royal Blue 4.7 mil Sustainable Nitrile Exam Gloves
Fournisseur: SW Safety Solutions
Certificats
A propos de cet article
Economical and high-performance hand protection for lab, life science, and industrial environments.
- EcoTek® sustainable technology
- Textured surface for excellent wet and dry grip
- Silicone-free for critical environments
- Tear-resistant beaded cuff eases donning
- SHA dermatologically approved
- 1.5 AQL exceeds laboratory standards
Value and performance hand protection for laboratory, life science, and industrial environments. EcoTek® gloves tested to ASTM D5526-12 standard test method for determining anaerobic biodegradation of plastic materials under accelerated landfill conditions. Results are 92.6% biodegradation in 60% solids landfill at 945 days. Future results cannot be predicted/extrapolated. For additional information on the test reports, contact [email protected].
Certifications: WBENC Women Owned; NCMSDC Minority Business; Small Business Enterprise; SHA Dermatologically Approved; EAC TP TC 019/2011; CE 0493; CE EN 455.
Ordering information: Glove sizes are as follows; XS (5.5 to 6), S (6.5 to 7), M (7.5 to 8), L (8.5 to 9), XL (9.5 to 10).
Spécifications
- Catalog No:
- 76382-948
- 76382-950
- 76382-944
- 76382-946
- 76382-954
- 76382-952
- Model:
- TF-95RB
- TF-95RB
- TF-95RB
- TF-95RB
- TF-95RB
- TF-95RB
- Ambidextrous/Hand-specific:
- Ambidextrous
- Ambidextrous
- Ambidextrous
- Ambidextrous
- Ambidextrous
- Ambidextrous
- Disposable / reusable:
- Disposable
- Disposable
- Disposable
- Disposable
- Disposable
- Disposable
- Latex/Latex-free:
- Latex-free
- Latex-free
- Latex-free
- Latex-free
- Latex-free
- Latex-free
- Powdered/Powder-free:
- Powder-free
- Powder-free
- Powder-free
- Powder-free
- Powder-free
- Powder-free
- EN norm:
- EN ISO 21420:2020; EN 455-2000-2015
- EN ISO 21420:2020; EN 455-2000-2015
- EN ISO 21420:2020; EN 455-2000-2015
- EN ISO 21420:2020; EN 455-2000-2015
- EN ISO 21420:2020; EN 455-2000-2015
- EN ISO 21420:2020; EN 455-2000-2015
- Food contact approved:
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes