Avantor® Hichrom, HPLC Column Connectors
# de catalogue 76399-058
Fournisseur: HICHROM LIMITED
Spécifications
- I.D.:0.010"
- Length:40 mm
- O.D.:¹/₁₆"
- Description:HPLC column connector
- Material:Stainless steel
- Cat. no.:76399-058
Spécifications
A propos de cet article
These column connectors are used to connect Hichrom guard and analytical columns.
- Stainless steel connector
- Female-female end fittings
The connector consists of a stainless steel tubing with appropriate 1/16" nuts and ferrules and can be used to connect female end fitting Hichrom guard and analytical columns and make similar connections.
Customers Who Bought This Also Bought