Avantor® Hichrom, Chromatography Tubing, Stainless Steel
Fournisseur: HICHROM LIMITED
Certificats
Certificats
A propos de cet article
Tubings are made of 316 stainless steel.
- Resistant to chemicals
The precision bore is ¹/₁₆" type tubing.
Customers Who Bought This Also Bought
VWR® Screw Caps for 9 mm Screw-Thread Vials, Wide Opening
Fournisseur: VWR International
57,65 $ to 603,92 $
VWR® 9 mm Screw-Thread Vials, Kits, Caps, and Inserts, Wide Opening
Fournisseur: VWR International
45,27 $
SureSTART™ Glass Crimp Top Headspace Vials, Level 3 High Performance Applications, Thermo Scientific
Fournisseur: Thermo Scientific
72,92 $ to 78,40 $
SureSTART™ Septa for 20 mm Stoppers, Level 2 High-throughput Applications, Thermo Scientific
Fournisseur: Thermo Scientific
131,41 $ to 136,31 $
WHEATON® MicroLiter 9 mm Screw-Thread Vials, DWK Life Sciences
Fournisseur: DWK Life Sciences (KIMBLE)
132,33 $
WHEATON® MicroLiter 20 mm Crimp Seals with Septa, DWK Life Sciences
Fournisseur: DWK Life Sciences (KIMBLE)
160,41 $
WHEATON® MicroLiter µLPlate® Component Kits, Inserts and Caps, Unassembled, DWK Life Sciences
Fournisseur: DWK Life Sciences (KIMBLE)
224,41 $