Avantor® Hichrom, Preparative Frit Assembly
# de catalogue 76399-164
Fournisseur: HICHROM LIMITED
Spécifications
- Porosity:2.0 µm
- Description:Preparative frit assembly, natural
Spécifications
A propos de cet article
Customers Who Bought This Also Bought
VWR® 9 mm Screw-Thread Vials, Kits, Caps, and Inserts, Wide Opening
Fournisseur: VWR International
$50.14 to $501.54
Sodium hydroxide 0.1 N in aqueous solution, VWR Chemicals BDH®
Fournisseur: VWR International
$194.14 to $694.12
VWR® Screw Caps for 9 mm Screw-Thread Vials, Wide Opening
Fournisseur: VWR International
$46.59 to $465.92
AQ™ Advanced Quality Borosilicate Glass and Plastic Inserts
Fournisseur: MicroSolv Technology Corp
$152.87
WHEATON® MicroLiter Sample Vial Component Kits, DWK Life Sciences
Fournisseur: DWK Life Sciences (KIMBLE)
$175.49
RSA™ and RSA-Pro™ Autosampler Vials, Caps and Convenient Kits
Fournisseur: MicroSolv Technology Corp
$438.81
SureSTART™ Screw Caps, 9 mm, Level 3 High Performance Applications
Fournisseur: Thermo Scientific
$79.21 to $82.14