Exsil Silica, HPLC Columns
Fournisseur: Avantor
Certificats
A propos de cet article
Exsil HPLC columns are a useful alternative to the classical spherical HPLC silicas.
- Spherical porous silica
- 80 and 100 Å pore sizes
- Competitively priced
- Hichrom high efficiency
Exsil 100, the standard material, is supplied in a range of particle sizes and bonded phases to cover a wide range of applications. Exsil 80 is a higher surface area, complementary range of materials.
Spécifications
- Catalog No:
- 76398-618
- Surface area:
- 200 m²/g