VeriSpec® ICP Multi-Element Standard 4, RICCA Chemical Company
Fournisseur: Ricca Chemical
Certificats
A propos de cet article
4 component standard.
5000 ppm: Ca, K, Mg, Na
Certifications: Manufactured and Tested in an ISO 17025/ISO 17034 Accredited Facility
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.