LiChrospher® RP-8E, HPLC Columns
Fournisseur: HICHROM LIMITED
Certificats
A propos de cet article
LiChrospher® is a silica available in both endcapped and non-endcapped C8 and C18 phases.
- Spherical porous silica
- High efficiency packed columns
Spécifications
- Catalog No:
- 76400-716
- 76400-704
- Carbon load:
- 13.0%
- 13.0%
- Endcapped:
- Yes
- Yes
- Surface area:
- 350 m²/g
- 350 m²/g