Avantor® Hichrom, UHPLC Column Connector
# de catalogue 76399-754
Fournisseur: HICHROM LIMITED
Spécifications
- Description:UHPLC connector for 1/₁₆" O.D. tubing
- Cat. no.:76399-754
Spécifications
A propos de cet article
This reusable UHPLC column connector have 10-32 threads. This fitting is reusable, as they are not permanently swaged onto the inlet tubing.
- Compatible with all UHPLC systems and column brands
- Avoids poor connections
- Pressure rated to >1700 bar (>25000 psi)
An internal ferrule design prevents tubing slippage, whilst the PEEK-polymer blend front ferrule eliminates the risk of damage to receiving ports.
Customers Who Bought This Also Bought