LiChrospher® RP-8, HPLC Columns
Fournisseur: HICHROM LIMITED
Certificats
A propos de cet article
LiChrospher® is a silica available in both endcapped and non-endcapped C8 and C18 phases.
- Spherical porous silica
- High efficiency packed columns
- Stainless steel column hardware
Spécifications
- Catalog No:
- 76400-650
- 76400-652
- 76400-654
- 76400-666
- 76400-668
- 76400-656
- 76400-680
- Carbon load:
- 12.5%
- 12.5%
- 12.5%
- 12.5%
- 12.5%
- 12.5%
- 12.5%
- Endcapped:
- No
- No
- No
- No
- No
- No
- No
- Surface area:
- 350 m²/g
- 350 m²/g
- 350 m²/g
- 350 m²/g
- 350 m²/g
- 350 m²/g
- 350 m²/g