Avantor® Hichrom, Chromatography Tubing, PEEK
Fournisseur: HICHROM LIMITED
Certificats
A propos de cet article
PEEK (polyetheretherketone) polymer tubing is biocompatible and chemically inert to most solvents. It is completely flexible, can easily be cut to desired lengths and is suitable for a wide range of applications.
- Biocompatible, inert and easily cut
- Good for high pressure applications
- Can be used with stainless steel or polymer fittings
- ¹/₁₆" O.D. tubing
It has a very smooth internal surface and is the least permeable to gas of all the polymer tubing materials. Alternative O.D. tubing, example ¹/₈" for high pressure semi-preparative systems and 360 μm for capillary systems, is also available.
Customers Who Bought This Also Bought