Avantor® Hichrom, HPLC Columns, Zinc
Fournisseur: HICHROM LIMITED
Certificats
Customers Who Bought This Also Bought
VWR® Snap Caps for 11 mm Snap Ring Vials, Wide Opening
Fournisseur: VWR International
73,75 $ to 568,62 $
VWR® Screw Caps for 8-425 Screw-Thread Vials, Standard Opening
Fournisseur: VWR International
120,75 $
WHEATON® MicroLiter Screw-Thread Headspace Vials, 16 mm, DWK Life Sciences
Fournisseur: DWK Life Sciences (KIMBLE)
139,00 $
WHEATON® MicroLiter 9 mm Screw-Thread Vials, Component Kits, Unassembled, DWK Life Sciences
Fournisseur: DWK Life Sciences (KIMBLE)
114,97 $
SureSTART™ Glass Screw Top Vials, 2 ml, Level 3 High Performance Applications, Thermo Scientific
Fournisseur: Thermo Scientific
48,65 $ to 52,28 $