Avantor® Hichrom, PEEK Tubing Elbows
Fournisseur: HICHROM LIMITED
Certificats
Certificats
A propos de cet article
Made up of PEEK material. These tubing offers perfect fit.
Customers Who Bought This Also Bought
VWR® Snap Caps for 11 mm Snap Ring Vials, Wide Opening
Fournisseur: VWR International
$73.75 to $568.62
VWR® Screw Caps for 9 mm Screw-Thread Vials, Wide Opening
Fournisseur: VWR International
$57.65 to $603.92
WHEATON® MicroLiter 9 mm Screw-Thread Vials, Component Kits, Unassembled, DWK Life Sciences
Fournisseur: DWK Life Sciences (KIMBLE)
$81.80
WHEATON® MicroLiter Septa for 20 mm Crimp Seals, DWK Life Sciences
Fournisseur: DWK Life Sciences (KIMBLE)
$182.14
SureSTART™ GOLD-Grade Glass Insert for 2 ml Vials, Level 3 High Performance Applications, Thermo Scientific
Fournisseur: Thermo Scientific
$810.00 to $840.15
VWR® 11 mm Crimp Top Vials, Kits, Caps, and Inserts, Wide Opening
Fournisseur: VWR International
$291.56
WHEATON® MicroLiter Sample Vial Component Kits, DWK Life Sciences
Fournisseur: DWK Life Sciences (KIMBLE)
$177.30
VWR® 9 mm Screw-Thread Vials, Kits, Caps, and Inserts, Wide Opening
Fournisseur: VWR International
$152.32
WHEATON® E-Z Vial® Large-Opening Crimp-Top Vials, DWK Life Sciences
Fournisseur: DWK Life Sciences (KIMBLE)
$110.04