Avantor® Hichrom, Chromatography Tubing, Stainless Steel
Fournisseur: HICHROM LIMITED
Certificats
A propos de cet article
Tubings are made of 316 stainless steel.
- Resistant to chemicals
The precision bore is ¹/₁₆" type tubing.
Customers Who Bought This Also Bought